Troubleshooting Guide - Tank Level Monitor Malfunction Alert - Aquanex Water Purification Systems
Description:
This article addresses troubleshooting a tank level monitor malfunction alert, focusing on scenarios where the tank level remains below 90% for over 10 minutes without volume increase. It details the process logic for RO water production, conditions for refilling, and communication issues with the Aquanex unit. Troubleshooting steps include reducing the water level, checking for water level percentage increases, and inspecting the feed water supply, pretreatment, RO membrane, or level sensor if the level does not rise. It emphasizes that Type I water cannot be dispensed while Type II water is being dispensed.
Environment:
Model 7201030 Aquanex 30L System with Dispenser and 30L tank
Model 7201031 Aquanex 30L System with Dispenser, 30L tank and Pretreatment
Attachment(s):
| File | Last Modified |
|---|---|
| Aquanex Level Monitor Malfuction alert V2.pdf | July 10, 2025 |