Calibration procedure for Bantam deionizer Model D0800
This article provides detailed instructions on calibrating the Model D0800 Bantam Deionizer using an external resistor. The calibration process involves setting the on/off switch to the "off" position, checking and adjusting the zero alignment of the meter needle, connecting a known resistance across the cell terminals, and adjusting the potentiometer so the meter reading corresponds to the simulated resistance. These steps ensure accurate calibration and optimal performance of the deionizer.
Description:
Instructions to adjust the analog purity meter found on model D0800 Bantam cartridge holder. Requires the use of a 50K ohm resistor.
Environment:
Thermo Scientific Bantam Deionizer Type D0800, series 494, model numbers:
| Model number | Voltage |
| D0800 (7107) | 120V |
| D0805 (7108) | 240V |
Attachment(s):
| File | Last Modified |
|---|---|
| D0800 Bantam Deionizer Meter Check and Adjustment Rev 2.pdf | May 13, 2025 |