Thermo Scientific Connect Edge - Configuration Planning Guide
The Connect Edge Network Configuration Planning Guide provides instructions for setting up and connecting Thermo Fisher's Connect Edge system to monitor laboratory devices. It covers data collection methods, network configuration options, and steps for connecting gateways and adapters to the Internet or local networks. The guide includes details on network connection types, device listing, and considerations for multiple gateways and network requirements.
Description:
This document is to provide guidance, configuration, and strategy setup with Connect Edge.
Environment:
Connect Edge, guidance, configuration, setup, thermo connect
| Connect Edge GateWay | Connect Edge Gateway Kit | Connect Edge Adapter Kits | Supported Equipment |
| EDGEPTGATE01 | EDGEKTETHE01 | EDGEKTADPT01 | Incubator |
| EDGEPTGATE02 | EDGEKTWIFI01 | EDGEKTADPT02 | Heracell Series |
| EDGEPTGATE03 | EDGEKTCELL01 | EDGEKTADPT03 | EDGEKTADPT03 (RS232 configuration) & EDGEKTADPT53 (RS232 configuration) |
| EDGEPTGATE04 | EDGEKTCELL02 | EDGEKTADPT04 | Heracell VIOS Series E |
| EDGEPTGATE05 | EDGEKTCELL03 | EDGEKTADPT51 | DGEKTADPT04 (USB configuration) & EDGEKTADPT54 (USB configuration) |
| EDGEPTGATE06 | EDGEKTCELL04 | EDGEKTADPT52 | ULT Freezers |
| EDGEPTGATE07 | EDGEKTCELL05 | EDGEKTADPT53 | UxF/HFU-T/TSU/88000, TSX Series, & TDE/RDE/FDE/HDE Series |
| EDGEPTGATE08 | EDGEKTCELL06 | EDGEKTADPT54 | EDGEKTADPT01 (RS485 configuration) & EDGEKTADPT51 (RS485 TSX ULT Series configuration) |
| EDGEKTADPT99 | TSX HPLRF | ||
| Freezer TSX Freezer Series EDGEKTADPT02 (STTL configuration) | |||
| Refrigerator TSX Refrigerator Series EDGEKTADPT52 (STTL configuration) | |||
| Refrigerator TSX Refrigerator Series EDGEKTADPT52 (STTL configuration) |
Attachment(s):
| File | Last Modified |
|---|---|
| Connect Edge Configuration Planning Rev B Dec 2024.pdf | January 20, 2025 |
| Thermo Scientific Connect Edge - Configuration Planning Guide.pdf | August 08, 2024 |