DeviceLink Connect - How to setup multidrop when using multiple sensors on an incubator
This article guides on setting up a Device Link Connect (DLC) for multiple sensors on an incubator. It details configuring a 90% humidity probe and a CO2 probe using a multidrop setup, assigning unique addresses, and connecting both sensors to a single RS232/RS485 port via a splitter. Users must then configure and deploy the settings to save changes.
Description:
Instructions on how to setup or install DeviceLink multidrop when using multiple sensors on an incubator.
Environment:
Directions, installation, Heracell VIOS, DLC, Remote Monitoring
Forma i160, Forma i250, and Forma Series 3 CO2 incubators (USB) DLCKTCO2I102
Heracell CO2 incubator (serial) DLCKTCO2I101,
Splitter connector cable This cable enables the connection of both CO₂ and temp-RH sensors to a single mote via the RS232/RS485 connection port DLCPTSPLI001
Attachment(s):
| File | Last Modified |
|---|---|
| DeviceLink Connect - How to Set up a Multidrop.pdf | September 22, 2023 |