Vios Incubator - How to set up the Telemetry Cable Set Instruction Sheet
This instruction sheet details the process for setting up the telemetry cable for Vios Incubators. It includes changing the BAUD rate to 9600 on both the incubator and DLC, with specific steps for accessing and adjusting the interface settings. Users must add sensor details, including type, USB port, baud rate, and voltage, then submit this information. Additionally, they need to enter their account email and device serial number before deploying changes to save the settings.
Description:
How to set up the Telemetry Cable Set for Vios Incubator to use with devicelink connect. Linking to instrument health via DLC requires Baud rate settings change on incubator.
Environment:
DLC, DeviceLink connect, Instrument Health, data push smart cable Vios incubator, Telemetry Cable 328928H02 (DLCPTDATA104) 160i /250i
Attachment(s):
| File | Last Modified |
|---|---|
| Vios Incubator - How to set up the Telemetry Cable Set Instruction Sheet.pdf | April 26, 2023 |