Smart-Tracker - How to Collect Detailed Log Files
This instruction sheet explains how to collect detailed log files from a Smart-Tracker device using the Smart-Vue Classic software. The process involves selecting the Settings tab, ensuring detailed logs are enabled, reconnecting to the Smart Tracker module, and copying the log data from the History tab.
Description:
Collecting detailed log files from a Smart-Tracker device
Environment:
Smart-Tracker
Attachment(s):
| File | Last Modified |
|---|---|
| Rev 2 - Smart-Tracker - How to Collect Detailed Log Files.pdf | January 29, 2022 |
| Smart-Tracker - How to Collect Detailed Log Files.pdf | January 29, 2022 |