Thermo Fisher Scientific
Smart Connect Mobile Application - Technical Data Sheet
This technical data sheet outlines the Smart Connect mobile app by Thermo Fisher Scientific, which enables control and data reading from Smart-Vue Pro Quatro/Duo data loggers via smartphones or tablets. It supports data push to the Cloud and alarm notifications via call, SMS, and email. Compatible with Android (versions 6-10) and iOS (versions 11-13.4), the app offers data export formats, local storage, graphical and tabular data viewing, alarm indications, and user authentication modes.
Description: 
Smart Connect mobile application is used to control and read Smart-Vue Pro Quatro/Duo data loggers from a phone or tablet
Environment:
Smart Connect mobile application for:
  • Smart-Vue Pro Quatro
  • Smart-Vue Pro Duo
  • Was this article helpful?