TCP and IP Power over Ethernet Receiver - Technical Data Sheet Rev Sep 2014
This technical data sheet outlines the TCP/IP POE Receiver by Thermo Fisher Scientific. It measures 190 x 52 x 30 mm, weighs 145 g, and supports 100 - 240 V AC to 5V DC power. It offers multiple mounting options and a 1-year warranty. Operating on ISM band frequencies, it has a wireless range up to 700 meters in open spaces. The receiver includes various alarms, scalable software, and FCC Part 15 compliance. It works within 0°C to 50°C, up to 90% RH non-condensing, and features IP65 protection.
Description:
Technical data sheet
Environment:
Thermo Scientific Smart-Vue Wireless Monitoring Solution, TCP/IP POE (Power over Ethernet) Receiver
Attachment(s):
| File | Last Modified |
|---|---|
| Rev 09_17_2014 - TCP_IP POE (Power over Ethernet) Receiver - Technical Data Sheet.pdf | January 29, 2022 |