Thermo Scientific - Smart Siren - Technical Data Sheet
This technical data sheet describes the Smart Siren. It provides visual and audio alerts for alarm conditions in the Smart-Vue Pro monitoring system. Designed for indoor use, it operates in temperatures from 0°C to +50°C and 0% to 90% RH. It can be mounted with magnets or Velcro® tape and is powered by an AC/DC adapter with extended battery life. The device supports LoRaWAN communication and is certified by multiple agencies. It includes a one-year warranty.
Description:
Technical data sheet
Environment:
- Smart siren 915 - SVPACCSIR001
- Smart siren 865-868 - SVPACCSIR002
Attachment(s):
| File | Last Modified |
|---|---|
| Rev A - Smart Siren - Technical Data Sheet.pdf | January 29, 2022 |