Smart-Vue Pro Wireless Monitoring Solution - 3.5 Meter Digital Sensor -40C to 80C - Technical Data Sheet Rev A
This technical data sheet provides details on the Smart-Vue Pro Wireless Monitoring Solution's 3.5 Meter Digital Sensor, catalog numbers SVPSENTUE001 and SVPSENTCE001. These sensors are used for monitoring conditions in refrigerators, freezers, ovens, and incubators, with a temperature range of -40°C to +80°C, measurement uncertainty of ±0.06°C to 0.25°C, and IP67 protection. The sensors are calibrated at specific temperatures and comply with ROHS and CE standards, offering a 1-year warranty.
Description:
Technical data sheet
Environment:
Smart-Vue Pro Wireless Monitoring Solution: 3.5 Meter Digital Sensor (-40°C to +80°C), catalog numbers:
- SVPSENTUE001
- SVPSENTCE001
Attachment(s):
| File | Last Modified |
|---|---|
| Rev A - Smart-Vue Pro Wireless Monitoring Solution_ 3.5 Meter Digital Sensor (-40C to +80C) - Technical Data Sheet.pdf | January 29, 2022 |