Smart-Vue Pro Wireless Monitoring Solution - 1 Meter Digital Sensor -40C to 80C - Technical Data Sheet Rev A
The article provides detailed specifications for the Smart-Vue Pro Wireless Monitoring Solution, specifically the 1 Meter Digital Sensor (Catalog Numbers: SVPSENTUS001, SVPSENTCS001). It outlines its application for monitoring various environments like refrigerators and incubators, its technical features including a temperature range of -40°C to +80°C, and accuracy within ±1°C. The sensor is made with PVC and stainless steel, is IP67 rated, and calibrated according to NIST and COFRAC standards.
Description:
Technical data sheet
Environment:
Smart-Vue Pro Wireless Monitoring Solution: 1 Meter Digital Sensor (-40°C to +80°C), catalog numbers:
- SVPSENTUS001
- SVPSENTCS001
Attachment(s):
| File | Last Modified |
|---|---|
| Rev A - Smart-Vue Pro Wireless Monitoring Solution_ 1 Meter Digital Sensor (-40C to +80C) - Technical Data Sheet.pdf | January 29, 2022 |