Smart-Vue Pro LoRaWAN Enabled Receiver - Technical Data Sheet
This technical data sheet outlines the Smart-Vue Pro LoRaWAN Enabled Receiver. It supports long-range wireless communication via LoRaWAN and connects through Ethernet or Wi-Fi. Designed for indoor use, it operates in temperatures from -30°C to +70°C and 20% to 90% RH. Features include an anodized aluminum casing, various security protocols, and browser support. It is certified by multiple agencies and includes a one-year warranty.
Description:
Technical data sheet
Environment:
Smart-Vue Pro LoRaWAN Enabled Receiver:
- Smart-Vue Pro LoraWAN 915 - SVPHWRLGW001
- Smart-Vue Pro LoraWAN 865-868 - SVPHWRLGW002
Attachment(s):
| File | Last Modified |
|---|---|
| Rev A - Smart-Vue Pro LoRaWAN Enabled Receiver - Technical Data Sheet.pdf | January 29, 2022 |