Smart-Vue Pro Wireless Monitoring Solution - 3.5 Meter PT100 Smart sensor -100C to 150C - Technical Data Sheet Rev A
This technical data sheet outlines the specs for the Smart-Vue Pro Wireless Monitoring Solution's 3.5 Meter PT100 Smart sensor, catalog numbers SVPSENTUS003 and SVPSENTCS003. Designed for monitoring ultra-low temperature freezers, the sensor measures temperatures from -100°C to +150°C with measurement uncertainty of ±0.08°C to ±0.3°C and accuracy of ±1°C. It features a 3.5-meter PVC cable, IP66 protection, and complies with ROHS and CE standards. Calibration is performed at -80°C, -40°C, and 0°C
Description:
Technical data sheet
Environment:
Smart-Vue Pro Wireless Monitoring Solution: 3.5 Meter PT100 Smart sensor (-100°C to +150°C), catalog numbers
- SVPSENTUS003
- SVPSENTCS003
Attachment(s):
| File | Last Modified |
|---|---|
| Rev A - Smart-Vue Pro Wireless Monitoring Solution_ 3.5 Meter PT100 Smart sensor (-100C to +150C) - Technical Data Sheet.pdf | January 29, 2022 |