Smart-Vue Pro Wireless Monitoring Solution - Temp and RH Smart sensor - Technical Data Sheet Rev A
This technical data sheet details the Smart-Vue Pro Wireless Monitoring Solution's Temp/RH Smart sensor (SVPSENRUS001, SVPSENRCS001, SVPSENRCS002). Designed for monitoring storage facilities and stability chambers, the sensor measures -40°C to +150°C and 0 to 99.9% RH, with ±0.3°C to ±0.5°C and ±4% RH measurement uncertainty. It features IP65 protection, ROHS and CE compliance, and a 1-year warranty.
Description:
Technical data sheet
Environment:
Smart-Vue Pro Wireless Monitoring Solution: Temp/RH Smart sensor (-40°C to +150°C, 0 to 99.9% RH), catalog numbers:
- SVPSENRUS001
- SVPSENRCS001
- SVPSENRCS002
Attachment(s):
| File | Last Modified |
|---|---|
| Rev A - Smart-Vue Pro Wireless Monitoring Solution Temp_RH Smart sensor - Technical Data Sheet.pdf | January 29, 2022 |