Smart Connect Mobile Application - Technical Data Sheet
This technical data sheet for the Smart Connect Mobile Application by Thermo Fisher Scientific details its specifications and features. The app controls and monitors Smart-Vue Pro Quatro/Duo data loggers via smartphones or tablets, with cloud data upload and alarm notifications. It supports Android (versions 6–10) and iOS (versions 11–13.4), requires permissions for Bluetooth, geolocation, internet connectivity, and camera access, and offers data export in PDF, Excel, and CSV formats.
Description:
Smart Connect mobile application is used to control and read Smart-Vue Pro Quatro/Duo data loggers from a phone or tablet
Environment:
Smart Connect mobile application for:
- Smart-Vue Pro Quatro
- Smart-Vue Pro Duo
Attachment(s):
| File | Last Modified |
|---|---|
| Rev A - Smart Connect Mobile Application - Technical Data Sheet.pdf | January 29, 2022 |