Smart Siren - Technical Data Sheet
This technical data sheet outlines the Smart Siren by Thermo Fisher Scientific. It details specifications, features, and usage. The Smart Siren is an indoor device with a temperature range of 0°C to +50°C and a humidity range of 0 to 90% RH. It uses a 12V DC power adapter and includes a lithium battery. The device employs LoRaWAN for connectivity and provides visual and audio alarms. It meets various standards and is available in North America and Europe.
Description:
Technical data sheet
Environment:
- Smart siren 915 - SVPACCSIR001
- Smart siren 865-868 - SVPACCSIR002
Attachment(s):
| File | Last Modified |
|---|---|
| Rev A - Smart Siren - Technical Data Sheet.pdf | September 15, 2023 |