AC/DC Shelf Replacement - DT00301 - Rev A - Field Repair
Issue
•Powering up the Bigfoot fails
•Info, Voltage BP 24.V-1 Out of Spec
•Info, Voltage BP 24.V-2 Out of Spec
•VoltageBpP240VA or VoltageBpP240VB will not be close to 24V (seen in the Status and Control Panel)
•The processing chassis and backplane fans do not have power and are unable to boot
•FPGAs are not powered
Environment
All Bigfoot Spectral Cell Sorter Models
Resolution
See attachment for complete resolution
Attachment(s)
File | Last Modified |
---|---|
DT00301-ACDC Shelf Replacement - Field Repair.pdf | July 05, 2023 |