Beam Width Sweep Standalone Procedure - DT00353 - Rev A
Issue
•Frequent re-alignment of 1x or more lasers
•Large Instability when subjected to environmental temperature changes
•Consistently has to go into MESH and/or Sub-MESH to find the best alignment position
•Higher than typical PMT Voltages on CellX2 or UV Lasers
Environment
All Bigfoot Spectral Cell Sorter Models
Resolution
See attachment for complete resolution
Attachment(s)
File | Last Modified |
---|---|
DT00353 - Beam Width Sweep Procedure.pdf | April 04, 2024 |