Detection and Bulk Fluids Door Hall Effect Sensor Replacement - DT00238 - Rev A - Field Repair
Issue
Detection Door
•Failure in ‘Always Open’ and ‘Always Closed’ State - Filter Validation would not run during protocol after detection door has been opened and closed
Bulk Fluids Door
•Failure in ‘Always Open’ State – Blue bulk fluid indicator lights would always be on, even when door is closed
•Failure in ‘Always Closed’ State – Blue bulk fluid indicator lights would never show which set of tanks was active
Environment
All Bigfoot Spectral Cell Sorters
Resolution
See attachment for complete resolution
Attachment(s)
File | Last Modified |
---|---|
DT00238 - Field Repair - Replace Hall Effect Sensor, Detection Door & Bulk Fluids Door.pdf | May 17, 2023 |