Nozzle Fill with Ethanol - DT00240 - Rev A - Field Troubleshooting
Issue
Instrument is struggling to get into Maintain, stay in Maintain, experiencing spraying, or having side stream formation abnormalities. Perform this process prior to altering Frequency. You may also want to perform this process whenever the nozzle has been dry ie starting up a drained instrument or after swapping a nozzle body.
Environment
All Bigfoot Spectral Cell Sorters
Resolution
See attachment for complete resolution
Attachment(s)
File | Last Modified |
---|---|
DT00240 - Field Troubleshooting - Nozzle Fill with Ethanol.pdf | May 03, 2023 |