Streams Camera Focus Repair
Issue
Camera focus issues
Environment
Streams camera
Resolution
Follow these steps as needed to repair the Streams camera focus. These steps are described in detail in the attached document.
- Remove back cover
- Reset camera settings
- Make side streams
- Open iris all the way
- Adjust focus
- Adjust iris
- Adjust illumination
- Adjust exclusion zone
- Test focus, iris, and illumination
- Replace back cover
Attachment(s)
File | Last Modified |
---|---|
DT00215 - Streams Camera - Repair - Adjust Streams Camera Focus, Iris, Illumination.pdf | April 06, 2023 |