Swap It – GCMS Interface for ISQ GC Series and TSQ GC Series Mass Spectrometers - User Manual
Description:
Thermo Swap It GC / MS-interface enables the operator to switch easily
capillary columns without using any tools and without venting the
MS detector. Due to the ultra small ID of the Swap It transfer tube, the
entrance of air and air moisture into the MS is reduced to an absolute
minimum. After column exchange the system is ready for operation
within less than 5 minutes. Swap It avoids unscheduled disruptions in
your analytical work and highly improves laboratory productivity. The
complete system kit includes the parts listed below.
Environment:
ISQ GC Series
TSQ GC Series
Attachment(s):
File | Last Modified |
---|---|
ISQ_TSQ_8000_Swap_It_Manual_2012-03.pdf | March 31, 2025 |