PSB GCMS_2018_0001 TSQ 4.0 Unfiltered Signal
Description:
TSQ 4.0 includes feature to allow unfiltered signal to be acquired which may resolve issues with intermittent loss of signal for low intensity analytes.
Environment:
TSQ GC Series
Attachment(s):
| File | Last Modified |
|---|---|
| PSB GCMS_2018_0001 TSQ 4.0 Unfiltered Signal.pdf | July 01, 2024 |