Error Setting Nebulizer Value in Qtegra
Issue
During startup sequence, plasma goes out and error message "Error Setting Nebulizer Value" is returned in Log View
Environment
- iCAP Q
- iCAP RQ
- iCAP TQ
Resolution
Power cycle the instrument by shutting off the circuit breaker on the left hand side of the instrument and waiting 30 seconds before powering back on.