Loss of communication between iCAP Q and computer
Issue
How do I turn off the plasma on the iCAP Q if I lose communication between the instrument and the computer?
Environment
iCAP Q
Resolution
The best way to extinguish the plasma safely is to turn the torch box door handle. This will activate an interlock causing the plasma and instrument to shut down safely.