Red Low Exhaust Gas Flow Indicator
Issue
The Low Exhaust Gas Flow indicator is red when the exhaust flow is less than 0.1 mbar or a specified threshold.
Environment
iCAP Qnova Series ICP-MS
Resolution
- Check the exhaust hoses at the top of the instrument.
- Verify that the exhaust system on site is working properly.
NOTE: Your facility may have a separate control switch or valve to activate the exhaust.