Thermo Fisher Scientific
Strontium Interference on Calcium
Issue
Recovery of quality control and unknown samples could be excessively low when strontium-88 and calcium-44 are used during calibration.
Environment
  • iCAP Q
  • iCAP RQ
  • iCAP TQ
Resolution
If it is necessary to analyze calcium at a mass/charge of 44, then an interference equation can be calculated by determining the strontium overlap onto calcium using a single element strontium standard.
Cause
Isobaric overlap of double charge strontium-88 with calcium-44
  • Was this article helpful?