Thermo Fisher Scientific
FC - 2020.009 - Rev A - FAIMS diagnostic tests might be absent with Exploris 2.0 software
Description: 
For Instruments running Exploris 2.0 software, the TNG Tune Diagnostics pane does not immediately recognize the presence of FAIMS Pro and the FAIMS diagnostic tests might not appear in the Diagnostics pane.
Environment:
  • Faims Pro
  • Exploris
  • Was this article helpful?