Thermo Fisher Scientific
FC - 2021.005 - Rev A - FAIMS Carrier Gas setting
Description: 
FAIMS requires a carrier gas to desolvate ions. For most applications, the default flow rate is the appropriate flow rate. However, there might be an instance where the default flow rate is not the optimal flow rate. The ‘Total Carrier Gas Flow’ parameter allows the flow rate to be lowered for those instances where are lower flow rate is optimal for spray stability.
Environment:
  • Faims Pro