Thermo Fisher Scientific
FAIMS Pro with Orbitrap Fusion and Orbitrap Fusion Lumos Mass Spectrometer - Training Manual Version 1.0
Description: 
The FAIMS Pro interface has gone through a complete redesign.  The new hardware comprises the main control box MCB, the RF coil box and electrode assembly mounted to a collar flange, which is attached to the MS inlet.  Finally there is a bundled cable connecting the two units. 
Environment:
FAIMS Pro with Orbitrap Fusion  and Orbitrap Fusion Lumos Mass Spectrometer
  • Was this article helpful?