Source settings for Calibration
Description:
Due to the lower flow rates used when infusing calibrant, the physical location of the needle tip should be closer than for work at analytical flow rates.
- About "1.2" front-to-back (slightly closer to 1 if spray shield is not being used)
- Halfway between L-M
- Centered left-and-right
See the attached document for pictures of proper placement.
Environment:
- TSQ-II
- OptaMax NG
- IonMax NG
Attachment(s):
File | Last Modified |
---|---|
Source settings for Calibration.png | May 04, 2022 |