XCALI-98072 - Rev A - TSQ Series II Instrument Control Application Installation Instructions
Description:
This article provides Instrument control application installation instructions.
NOTE: The TSQ Series II version 3.2 instrument control software is compatible with the following software packages that support various liquid chromatography (LC) instruments:
• Standard Instrument Integration (SII) for Xcalibur version 1.4 or later
• LC Devices version 3.2 or later
• Chromeleon Chromatography Data System (CDS) version 7.2.10
• Aria MX 2.4 or later
Environment:
- TSQ Altis™
- TSQ Quantis™
- TSQ Fortis™
- TSQ Quantiva™
- TSQ Endura™ mass spectrometers
Attachment(s):
File | Last Modified |
---|---|
XCALI-98072 - Rev A - TSQ Series II Instrument Control Application Installation Instructions | January 09, 2022 |