Shut Down Procedure for the TSQ Quantiva and Endura
Description:
Shut down the TSQ Endura or TSQ Quantiva system completely only when you are not using it for an extended period of time or when you must shut it down for maintenance or service. You do not need to shut down the system completely if you are not going to use it temporarily, such as overnight or through the weekend. Instead, place the system in standby mode.
Environment:
- TSQ Quantiva system
- TSQ Endura system
Attachment(s):
File | Last Modified |
---|---|
Shut Down Procedure for the TSQ Quantiva and Endura.PDF | December 30, 2021 |