Thermo Fisher Scientific
Connection to Exploris can be lost if DNS is set to 127.0.0.0 on Network Card (NIC)
Issue
Connection / communication to Exploris can be lost if Preferred DNS Server is set to 127.0.0.0 on Network Card (NIC).
Environment
Exploris Series
Might be applicable to other instrument series
Resolution
Customer had Preferred DNS Server set to 127.0.0.0
clipboard_e1751f145c5420530cb48122ba41c0c55.png
Removed 127.0.0.0 and connection restored.
clipboard_e5db44f3d18ce9fb1dd8ea4d610027564.png
  • Was this article helpful?