3.5 KV patches for Electron Multipliers
Issue
How to check to have the 3.5 KV Dynode power supply installed. ( Information can be find in the instrument console during reboot )
Environment
- Velos
- Velos Pro
- Tune 2.7 sp2
- Orbitrap Elite
Resolution
- Two files need to be replaced. (init.ICB, detmult.ICB)
- After installation of the 2 files, reboot the system.
- The Electron Multiplier will be abe to go to 3500 Volts during calibration.