Thermo Fisher Scientific
Air leak
Issue
System air leak
Environment
Q Exactive GC
Resolution
Troubleshooting Tips:
  • Listen for a rush of air or a hissing sound inside the mass spectrometer. 
  • Possible causes of a major leak might be a loose or disconnected fitting, an improperly positioned O-ring, or an open valve.
  • If user suspects an air leak in the forevacuum region, check the vacuum tube for holes.
  • Check the clamp that fixes the vacuum tube to the forevacuum port (elbow) of the mass spectrometer. It might be loose because the forepump was shifted or the vacuum tube is twisted.
Cause
Sustained high pressure