Cleaning Agents
Issue
What are recommended cleaning agents for the instrument?
Environment
Q Exactive GC
Resolution
The following are recommended cleaning agents:
- A solvent like acetone (in accordance with your local safety practices).
- A detergent, for example, RBS 50 (trade name of Messrs. Carl Roth, Karlsruhe, Germany).
- Several liters of distilled water.