Deleting a Lock Mass Collection
Issue
Deleting a lock mass collection
Environment
Q Exactive GC
Resolution
- In the Lock Masses dialog box, select an existing lock mass collection in the list box.
- Click the button.
- In the Delete Lock Mass Collection dialog box, click OK to confirm your action and to close the dialog box.
- The name of the lock mass collection is removed from the list box of the Lock Masses dialog box.