How to Perform a NIST Search for a Mass Spectrum Using Chromeleon or NIST program Directly
Issue
Is there a way to search the GCMS NIST library for a specific compound in Chromeleon?
Environment
- Product model : ISQ, ISQ 7000, TSQ 8000, TSQ 9000
- Application: GC/GCMS, Chromeleon 7.2 and 7.3
Resolution
In order to find the Mass Spectrum of a Compound in NIST and how to navigate to NIST through Chromeleon 7:
Go directly to the NIST program C:\Program Files (x86)\NISTMS\MSSEARCH then NISTMS.exe. your executable may be somewhere else, but should be in this location.
Alternatively, go through Chromeleon:
Go to Chromatogram. Right-click. Select Show Spectrum. | ![]() |
Right click on Spectrum and Go to Open Spectrum with NIST. | ![]() |
When NIST opens you can go to Search and then you can search by CAS or several other parameters. | ![]() |
Attachment(s)
File | Last Modified |
---|---|
Figure 3 - Show Spectrum.png | January 16, 2022 |
Figure 2 - Show Spectrum.png | January 16, 2022 |
Figure 1 - Show Spectrum.png | January 16, 2022 |