How to display the error log in Xcalibur Raw FIles
Issue
How do I find the error log file in Xcalibur?
Environment
- Xcalibur
- GCMS
- LCMS
Resolution
Each individual raw file has its own error log.
- Open up the raw file in Qual Browser (for the run that had no injection, for example).
- Change the view in one of the frames to “Error Log” or “Status” by
- Put your mouse cursor over the frame that is active (push pin icon “green”).
- Right-mouse clicking.
- In the list of choices, select “View” and “Error Log”.
In error log, you may have an indication of if there was a “Fake Injection” or anything else that happened.