ISQ continues to lose communication
Issue
ISQ continues to lose communication
Environment
- GCMS ISQ
- GCMS TSQ
Resolution
- Look at the LAN Cable to see if it is damaged.
- Locate the cable that is included in the MS Accessory, Ping the IP of the MS System directly with CMD.
- If needed, change the Lan connection port on the PC, if no communication possible Digital load or Instrument control board may be faulty.