MS Communication error
Issue
MS Communication error; ISQ keeps losing comms
Environment
GCMS (ISQ, TSQ)
Resolution
- Look for the LAN Cable and see if its damaged.
- Look for the right cable, included in the MS Accessory.
- Ping the IP of the MS System directly with CMD.
- Change the LAN connection port on the PC.
- When no communication possible, digital board or instrument control board is faulty.