Vacuum leak
Issue
Unit has experienced a vacuum leak
Environment
Q Exactive GC
Resolution
- For proper performance, your Q Exactive GC system must operate at acceptable vacuum levels.
- User should check the current pressure values in the Vacuum / Bakeout window of the Tune software window.
- To display the Tune software window, choose Start > Programs > Thermo Exactive Series > Tune. The Instrument status LED in the toolbar turns green when the vacuum levels of the instrument are sufficient for operating your Q Exactive GC system.
- In the Tasks panel of the Tune software window, click to display the Vacuum / Bakeout window. Green LEDs indicate that the readback values are sufficient for operating the Q Exactive GC mass spectrometer.
Vacuum/Bakeout Button |
Green : sufficient for operating |
- Compare the current values of the pressures in the vacuum manifold with the values listed in Table 1 below. If the current values are higher than normal, there might be an air leak.
- If the pressure in the Ultra High Vacuum region is high (above 1E-9 mbar), the instrument needs to be baked out for about 10 hours or more.
Attachment(s)
File | Last Modified |
---|---|
vacuum leak - image 2.png | January 11, 2022 |
vacuum leak - image 1.png | January 11, 2022 |
vacuum leak - image 3.png | January 11, 2022 |