031197 - Rev 06 - AminoTrap Column - Product Manual
Description:
The AminoTrap column is used to retain amino acids present in carbohydrate samples and thereby prevent them from
interfering with the carbohydrate analysis. This manual describes the use of the AminoTrap column.
Environment:
- 4 x 50 mm P/N 046122
- 3 x 30 mm P/N 060146
- 0.4 x 35 mm P/N 076200
Attachment(s):
| File | Last Modified |
|---|---|
| 031197 - Rev 06 - AminoTrap Column - Product Manual.PDF | December 29, 2021 |