Smart calibration
Description:
The mass calibrations are now scripted Smart. If for the standard mass calibration it cannot find the calibrant peaks within the tolerance range, it will start over with an automated two-point calibration, eFT calibrations, etc to result in a successful calibration.
Environment:
Mass Spectrometry
Attachment(s):
File | Last Modified |
---|---|
Smart calibration.DOCX | December 31, 2021 |