Obtaining DPPCal2 troubleshooting data from NS7 EDS install and calibration
Description:
Irregular results are seen when calibrating an EDS detecter on an NS7 system
Environment:
- Noran Ssytem 7
Resolution:
- NS7 has the ability to store-save-amend crucial DPP parameters in csv files for review by engineers at the factory to aid in resolving irregular calibration issues.
- Location of this data is in the Shared Docs folder in a separate "DPPCal Data" folder.