AN 51947 - Rapid Quality Screening of Carbon Nanotubes with Raman Spectroscopy
Carbon nanotubes are becoming available at commercially attractive prices, with a number of products incorporating nanotubes already available on the market today. As the quantities of carbon nanotubes being produced and used in commercial products ramps up, it becomes more and more important to have a quick means of verifying the quality of the material. Raman spectroscopy is one method that has become quite popular for this application. Raman measurements are fast and require minimal sample pr
Description:
Carbon nanotubes are becoming available at commercially attractive prices, with a number of products incorporating nanotubes already available on the market today. As the quantities of carbon nanotubes being produced and used in commercial products ramps up, it becomes more and more important to have a quick means of verifying the quality of the material. Raman spectroscopy is one method that has become quite popular for this application. Raman measurements are fast and require minimal sample preparation. hermo Scientific DXR Raman systems are ideally suited for this analysis because of their ease of operation and precise control of measurement parameters.
Environment:
DXR Raman
Attachment(s):
| File | Last Modified |
|---|---|
| AN51947 - Rapid Quality Screening of Carbon Nanotubes with Raman Spectroscopy.pdf | April 02, 2026 |