AN 52404 - Raman Spectroscopic Assessment of Purification Methods for Single Walled Carbon
Description:
Single walled carbon nanotubes hold tremendous promise in a variety of areas due to their unique physical and electronic properties. However, impurities can have a deleterious affect on these properties. This attached application note highlights how the DXR Raman microscope can be used to gauge the effectiveness of purification and enrichment methods.
Environment:
- DXR
Attachment(s):
| File | Last Modified |
|---|---|
| AN52404 - Raman Nanotubes Purification.pdf | June 02, 2023 |