TN1668 - Automated Sampling Array (ASA) for Multi-sample Measurements
Description:
This technical note describes the Automated Sampling Array (ASA) accessory for the Thermo Scientific DXR3 SmartRaman+ spectrometer, enabling automated, high-throughput Raman analysis of multiple samples.
It outlines system setup, software workflow, and optimization of sampling positions and focus, along with examples of customizable sample holders (e.g., vials, tablets, well plates, and bottles). The document highlights how ASA improves efficiency, flexibility, and reproducibility in applications such as pharmaceutical product qualification and quality assurance.
Note: The printed sample and vial holders shown are illustrative only and are not provided by Thermo Fisher Scientific. They demonstrate how users can design and customize holders to fit their specific applications.
Environment:
DXR3 SmartRaman+ only
(NOT for DXR3 SmartRaman)
Attachment(s):
| File | Last Modified |
|---|---|
| VIB NPI SmartRaman+ ASA tech note-MCS-TN1668-EN-12-2025-2865650-WEB.pdf | April 06, 2026 |