TN 1622 - Maintaining Optimum Raman Spectrometer Performance with the DXR3 SmartRaman Plus Alignment and Calibration Tool
This technical note describes the alignment and calibration workflow for the Thermo Scientific DXR3 SmartRaman+ spectrometer using the Alignment & Calibration Tool.
Description:
This technical note describes the alignment and calibration workflow for the Thermo Scientific DXR3 SmartRaman+ spectrometer using the Alignment & Calibration Tool. It explains automated procedures for instrument alignment, wavelength calibration, laser calibration, and relative intensity correction to ensure consistent, high-quality Raman results. The document highlights how these routines maintain optimal performance, improve spectral accuracy, and support reliable identification and quantitative analysis.
Note: The Laser Steer Assembly referenced is an internal instrument component used during alignment, not a user-accessible tool.
Environment:
DXR3 SmartRaman+ only
(NOT for DXR3 SmartRaman)
Attachment(s):
| File | Last Modified |
|---|---|
| SmartRaman+ calibration tech note-MCS-TN1622-EN-11-2025-2728850-WEB.pdf | April 06, 2026 |